Malonaldehyde bis(dimethyl acetal), 98%
Malonaldehyde bis(dimethyl acetal), 98%
Malonaldehyde bis(dimethyl acetal), 98%
Thermo Scientific Chemicals

Malonaldehyde bis(dimethyl acetal), 98%

CAS: 102-52-3 | C7H16O4 | 164.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 148611000
Price (USD)
38.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
38.00
Each
Chemical Identifiers
CAS102-52-3
IUPAC Name1,1,3,3-tetramethoxypropane
Molecular FormulaC7H16O4
InChI KeyXHTYQFMRBQUCPX-UHFFFAOYSA-N
SMILESCOC(CC(OC)OC)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4050 to 1.4090 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=97.5 %
Appearance (Color)Clear colorless to yellow-brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only