5-Methyl-2-hexanone, 99%
5-Methyl-2-hexanone, 99%
5-Methyl-2-hexanone, 99%
Thermo Scientific Chemicals

5-Methyl-2-hexanone, 99%

CAS: 110-12-3 | C7H14O | 114.19 g/mol
数量:
1 L
2.5 L
パッケージ:
Glass bottle
製品番号(カタログ番号) 149660025
価格(JPY)
-
数量:
2.5 L
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS110-12-3
IUPAC Name5-methylhexan-2-one
Molecular FormulaC7H14O
InChI KeyFFWSICBKRCICMR-UHFFFAOYSA-N
SMILESCC(C)CCC(C)=O
さらに表示
GC>=98.5 %
Refractive index1.4060 to 1.4090 (20°C, 589 nm)
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only