2-Methyl-2,4-pentanediol, 99%
2-Methyl-2,4-pentanediol, 99%
2-Methyl-2,4-pentanediol, 99%
Thermo Scientific Chemicals

2-Methyl-2,4-pentanediol, 99%

CAS: 107-41-5 | C6H14O2 | 118.18 g/mol
Quantity:
1 L
2.5 L
25 L
Packaging:
View all
Metal drum
Catalog number 150340250
Price (JPY)
-
Quantity:
25 L
Packaging:
Metal drum
Request bulk or custom format
Chemical Identifiers
CAS107-41-5
IUPAC Name2-methylpentane-2,4-diol
Molecular FormulaC6H14O2
InChI KeySVTBMSDMJJWYQN-UHFFFAOYNA-N
SMILESCC(O)CC(C)(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<15 APHA
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4260 to 1.4280 (20°C, 589 nm)
Appearance (Form)Clear liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only