1-Bromo-2-nitrobenzene, 98%
1-Bromo-2-nitrobenzene, 98%
1-Bromo-2-nitrobenzene, 98%
Thermo Scientific Chemicals

1-Bromo-2-nitrobenzene, 98%

CAS: 577-19-5 | C6H4BrNO2 | 202.01 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) 150350250
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS577-19-5
IUPAC Name1-bromo-2-nitrobenzene
Molecular FormulaC6H4BrNO2
InChI KeyORPVVAKYSXQCJI-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=CC=C1Br
さらに表示
GC>=97.5 %
Appearance (Color)Yellow
Appearance (Form)Fused solid or crystals and/or chunks
Infrared spectrumConforms
Melting point39°C to 43°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only