4-Butylaniline, 97%
4-Butylaniline, 97%
4-Butylaniline, 97%
Thermo Scientific Chemicals

4-Butylaniline, 97%

CAS: 104-13-2 | C10H15N | 149.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 150655000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS104-13-2
IUPAC Name4-butylaniline
Molecular FormulaC10H15N
InChI KeyOGIQUQKNJJTLSZ-UHFFFAOYSA-N
SMILESCCCCC1=CC=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to light brown
Appearance (Form)Liquid
GC>=96.0 %
Infrared spectrumConforms
Refractive index1.5340 to 1.5360 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only