Phenol Red, sodium salt, pure, water soluble, indicator
Phenol Red, sodium salt, pure, water soluble, indicator
Phenol Red, sodium salt, pure, water soluble, indicator
Thermo Scientific Chemicals

Phenol Red, sodium salt, pure, water soluble, indicator

CAS: 34487-61-1 | C19H13NaO5S | 376.358 g/mol
数量:
5 g
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 151441000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS34487-61-1
IUPAC Namesodium 4-[3-(4-hydroxyphenyl)-1,1-dioxo-3H-2,1λ⁶-benzoxathiol-3-yl]benzen-1-olate
Molecular FormulaC19H13NaO5S
InChI KeyHKHYOKBQJILTEI-UHFFFAOYNA-M
SMILES[Na+].OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C([O-])C=C1
さらに表示
Appearance (Form)Powder
Solubility(0.1 % in water) Clear
Infrared spectrumConforms
Visual transition intervalfrom pH 6.8 (yellow) to pH 8.2 (red)
Appearance (Color)Dark red to brown
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only