2,2,6,6-Tetramethyl-4-piperidinol, 99%
2,2,6,6-Tetramethyl-4-piperidinol, 99%
2,2,6,6-Tetramethyl-4-piperidinol, 99%
Acros Organics

2,2,6,6-Tetramethyl-4-piperidinol, 99%

CAS: 2403-88-5 | C9H19NO | 157.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
250 g
Packaging:
Glass bottle
Catalog number 151940250
also known as 15194
HSN 29333990
Price (INR)
15,465.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
15,465.00
Each
Chemical Identifiers
CAS2403-88-5
IUPAC Name2,2,6,6-tetramethylpiperidin-4-ol
Molecular FormulaC9H19NO
InChI KeyVDVUCLWJZJHFAV-UHFFFAOYSA-N
SMILESCC1(C)CC(O)CC(C)(C)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to off-white to light beige
Titration with HClO4>=98.5 %
Appearance (Form)Crystalline powder or crystals and/or chunks
Melting point129°C to 133°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only