2-Methyl-3-nitroaniline, 97%
2-Methyl-3-nitroaniline, 97%
2-Methyl-3-nitroaniline, 97%
Thermo Scientific Chemicals

2-Methyl-3-nitroaniline, 97%

CAS: 603-83-8 | C7H8N2O2 | 152.15 g/mol
数量:
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 152021000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS603-83-8
IUPAC Name2-methyl-3-nitroaniline
Molecular FormulaC7H8N2O2
InChI KeyHFCFJYRLBAANKN-UHFFFAOYSA-N
SMILESCC1=C(N)C=CC=C1[N+]([O-])=O
さらに表示
Infrared spectrumConforms
Melting point87°C to 91°C
Appearance (Form)Crystalline powder
HPLC>=96.0 %
Appearance (Color)Yellow to yellow-green
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only