3,3-Dimethylpentane, 99%
3,3-Dimethylpentane, 99%
3,3-Dimethylpentane, 99%
Thermo Scientific Chemicals

3,3-Dimethylpentane, 99%

CAS: 562-49-2 | C7H16 | 100.2 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
1 g
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
產品號碼 152660010
價格 (TWD)
-
Quantity:
1 g
Request bulk or custom format
化學識別
CAS562-49-2
IUPAC Name3,3-dimethylpentane
Molecular FormulaC7H16
InChI KeyAEXMKKGTQYQZCS-UHFFFAOYSA-N
SMILESCCC(C)(C)CC
檢視更多
規格Specification Sheet規格表
Appearance (Form)Liquid
GC>=98.5 %
Appearance (Color)Clear colorless
Refractive index1.3900 to 1.3920 (20°C, 589 nm)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only