Solketal, 97%
Solketal, 97%
Solketal, 97%
Thermo Scientific Chemicals

Solketal, 97%

CAS: 100-79-8 | C6H12O3 | 132.16 g/mol
数量:
100 g
500 g
2.5 kg
パッケージ:
Glass bottle
製品番号(カタログ番号) 153655000
価格(JPY)
-
数量:
500 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS100-79-8
IUPAC Name(2,2-dimethyl-1,3-dioxolan-4-yl)methanol
Molecular FormulaC6H12O3
InChI KeyRNVYQYLELCKWAN-UHFFFAOYNA-N
SMILESCC1(C)OCC(CO)O1
さらに表示
GC>=96.0 %
Refractive index1.4320 to 1.4360 (20°C, 589 nm)
Appearance (Color)Clear colorless
Infrared spectrumConforms
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only