2-Aminobenzyl alcohol, 98%
2-Aminobenzyl alcohol, 98%
2-Aminobenzyl alcohol, 98%
Thermo Scientific Chemicals

2-Aminobenzyl alcohol, 98%

CAS: 5344-90-1 | C7H9NO | 123.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 153731000
Price (USD)
188.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
188.00
Each
Chemical Identifiers
CAS5344-90-1
IUPAC Name(2-aminophenyl)methanol
Molecular FormulaC7H9NO
InChI KeyVYFOAVADNIHPTR-UHFFFAOYSA-N
SMILESNC1=CC=CC=C1CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point82°C to 86°C
Appearance (Color)Beige to grey-brown
GC>=97.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only