3-tert-Butylphenol, 99+%
3-tert-Butylphenol, 99+%
3-tert-Butylphenol, 99+%
Thermo Scientific Chemicals

3-tert-Butylphenol, 99+%

CAS: 585-34-2 | C10H14O | 150.22 g/mol
数量:
1 g
25 g
100 g
パッケージ:
すべて表示
Glass bottle
製品番号(カタログ番号) 154380010
価格(JPY)
-
数量:
1 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS585-34-2
Infrared spectrumConforms
GC>=99.0 %
Appearance (Form)Crystalline powder
Melting point40°C to 43°C
Appearance (Color)White to off-white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only