Diphenylcarbazone, suitable for colorimetric analysis
Diphenylcarbazone, suitable for colorimetric analysis
Diphenylcarbazone, suitable for colorimetric analysis
Thermo Scientific Chemicals

Diphenylcarbazone, suitable for colorimetric analysis

CAS: 538-62-5 | C13H12N4O | 240.26 g/mol
数量:
10 g
50 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 154450500
価格(JPY)
-
数量:
50 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS538-62-5
IUPAC Name3-(phenylamino)-1-(phenylimino)urea
Molecular FormulaC13H12N4O
InChI KeyZFWAHZCOKGWUIT-UHFFFAOYSA-N
SMILESO=C(NNC1=CC=CC=C1)N=NC1=CC=CC=C1
さらに表示
Appearance (Color)Orange to brown
Infrared spectrumConforms
Suitability(for Hg determination) Passes test
HPLC>=35 % (Diphenylcarbazone basis)
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only