Undecylenic acid, 98+%
Undecylenic acid, 98+%
Undecylenic acid, 98+%
Thermo Scientific Chemicals

Undecylenic acid, 98+%

CAS: 112-38-9 | C11H20O2 | 184.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 154495000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS112-38-9
IUPAC Nameundec-10-enoic acid
Molecular FormulaC11H20O2
InChI KeyFRPZMMHWLSIFAZ-UHFFFAOYSA-N
SMILESOC(=O)CCCCCCCCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Infrared spectrumConforms
GC>=97.0 %
Appearance (Form)Low melting solid or liquid
Titration with NaOH98.0 to 102.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only