UltraPure™ Sodium Dodecyl Sulfate (SDS) is a detergent that functions as a protein denaturant. It is used in the determination深入閱讀
Have Questions?
產品號碼
Quantity
15525017
500 g
產品號碼 15525017
價格 (TWD)
7,560.00
線上優惠
Ends: 30-Jun-2026
10,800.00
您節省 3,240.00 (30%)
Each
Quantity:
500 g
Request bulk or custom format
價格 (TWD)
7,560.00
線上優惠
Ends: 30-Jun-2026
10,800.00
您節省 3,240.00 (30%)
Each
UltraPure™ Sodium Dodecyl Sulfate (SDS) is a detergent that functions as a protein denaturant. It is used in the determination of protein molecular weight by polyacrylamide gel electrophoresis. It is also widely used to disrupt cell walls and dissociate nucleic acid-protein complexes in DNA and RNA extraction protocols.
For Research Use Only. Not for use in diagnostic procedures.
規格
For Use With (Application)Nucleic Acid Extraction
FormPowder
Quantity500 g
Reagent TypeSDS
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice