3-Aminobenzoic acid, 99+%
3-Aminobenzoic acid, 99+%
3-Aminobenzoic acid, 99+%
Thermo Scientific Chemicals

3-Aminobenzoic acid, 99+%

CAS: 99-05-8 | C7H7NO2 | 137.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 155850250
Price (EUR)
22,65
Online exclusive
24,60
Save 1,95 (8%)
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
22,65
Online exclusive
24,60
Save 1,95 (8%)
Each
Specifications
Assay Percent Range99+%
Beilstein14, 383
CAS Max %100.0
CAS Min %99.0
CAS99-05-8
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only