Indole-3-carboxaldehyde, 97%
Indole-3-carboxaldehyde, 97%
Indole-3-carboxaldehyde, 97%
Thermo Scientific Chemicals

Indole-3-carboxaldehyde, 97%

CAS: 487-89-8 | C9H7NO | 145.16 g/mol
数量:
5 g
25 g
100 g
パッケージ:
すべて表示
Plastic bottle
製品番号(カタログ番号) 156640250
価格(JPY)
-
数量:
25 g
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS487-89-8
IUPAC Name1H-indole-3-carbaldehyde
Molecular FormulaC9H7NO
InChI KeyOLNJUISKUQQNIM-UHFFFAOYSA-N
SMILESO=CC1=CNC2=CC=CC=C12
さらに表示
Infrared spectrumConforms
Melting point193°C to 199°C
Appearance (Color)Off-white to beige to light brown
Appearance (Form)Crystalline powder
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only