Maleimide, 98%
Maleimide, 98%
Maleimide, 98%
Thermo Scientific Chemicals

Maleimide, 98%

CAS: 541-59-3 | C8H4AgKN2O4 | 339.10 g/mol
数量:
5 g
10 g
25 g
100 g
500 g
パッケージ:
すべて表示
Glass bottle
製品番号(カタログ番号) 156710050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS541-59-3
IUPAC Namepotassium silver(1+) bis(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-ide)
Molecular FormulaC8H4AgKN2O4
InChI KeyKWLWLQRUFFGWNG-UHFFFAOYSA-L
SMILES[K+].[Ag+].O=C1[N-]C(=O)C=C1.O=C1[N-]C(=O)C=C1
さらに表示
GC>=97.5 %
Infrared spectrumConforms
Melting point90°C to 94°C
Titration with NaOH>=97.5 %
Appearance (Color)White to light yellow
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only