1-Methyl-4-piperidone, 98%
1-Methyl-4-piperidone, 98%
1-Methyl-4-piperidone, 98%
Thermo Scientific Chemicals

1-Methyl-4-piperidone, 98%

CAS: 1445-73-4 | C6H11NO | 113.16 g/mol
数量:
100 g
500 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 156925000
価格(JPY)
-
数量:
500 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1445-73-4
IUPAC Name1-methylpiperidin-4-one
Molecular FormulaC6H11NO
InChI KeyHUUPVABNAQUEJW-UHFFFAOYSA-N
SMILESCN1CCC(=O)CC1
さらに表示
Water=<0.5 % (K.F.)
Refractive index1.4590 to 1.4630 (20°C, 589 nm)
Appearance (Color)Clear yellow to orange to brown
Infrared spectrumConforms
GC>=97.5 %
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only