Uracil, 99+%
Uracil, 99+%
Uracil, 99+%
Thermo Scientific Chemicals

Uracil, 99+%

CAS: 66-22-8 | C4H4N2O2 | 112.09 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
產品號碼 157300025
價格 (TWD)
-
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
化學識別
CAS66-22-8
IUPAC Name1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular FormulaC4H4N2O2
InChI KeyISAKRJDGNUQOIC-UHFFFAOYSA-N
SMILESO=C1NC=CC(=O)N1
檢視更多
規格Specification Sheet規格表
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=99.0 %
Sulfated ash=<0.3 %
Heavy metals (as Pb)=<10 ppm
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only