Uracil, 99+%
Uracil, 99+%
Uracil, 99+%
Thermo Scientific Chemicals

Uracil, 99+%

CAS: 66-22-8 | C4H4N2O2 | 112.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 157301000
Price (EUR)
155,65
Online exclusive
173,00
Save 17,35 (10%)
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
155,65
Online exclusive
173,00
Save 17,35 (10%)
Each
Chemical Identifiers
CAS66-22-8
IUPAC Name1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular FormulaC4H4N2O2
InChI KeyISAKRJDGNUQOIC-UHFFFAOYSA-N
SMILESO=C1NC=CC(=O)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=99.0 %
Sulfated ash=<0.3 %
Appearance (Color)White to light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only