Pyridine-N-oxide, 95%
Pyridine-N-oxide, 95%
Pyridine-N-oxide, 95%
Thermo Scientific Chemicals

Pyridine-N-oxide, 95%

CAS: 694-59-7 | C5H5NO | 95.1 g/mol
数量:
25 g
100 g
500 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 157690250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS694-59-7
IUPAC Namepyridin-1-ium-1-olate
Molecular FormulaC5H5NO
InChI KeyILVXOBCQQYKLDS-UHFFFAOYSA-N
SMILES[O-][N+]1=CC=CC=C1
さらに表示
Water=<2 % (K.F.)
Appearance (Color)White to brown
Appearance (Form)Crystals or crystalline solid and/or chunks
Infrared spectrumConforms
Melting point61°C to 66°C
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only