2-(Methylamino)ethanol, 99%
2-(Methylamino)ethanol, 99%
2-(Methylamino)ethanol, 99%
Thermo Scientific Chemicals

2-(Methylamino)ethanol, 99%

CAS: 109-83-1 | C3H9NO | 75.11 g/mol
Quantity:
500 g
2.5 kg
10 kg
Packaging:
View all
Glass bottle
Catalog number 158260025
Price (JPY)
-
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS109-83-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=98.5 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4380 to 1.4400 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only