Glycerol, 99+%, Extra Pure
Glycerol, 99+%, Extra Pure
Glycerol, 99+%, Extra Pure
Thermo Scientific Chemicals

Glycerol, 99+%, Extra Pure

CAS: 56-81-5 | C3H8O3 | 92.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
10 L
25 L
Packaging:
View all
Glass Bottle
Catalog number 158920010
Price (EUR)
94,10
Each
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
94,10
Each
Chemical Identifiers
CAS56-81-5
IUPAC Namepropane-1,2,3-triol
Molecular FormulaC3H8O3
InChI KeyPEDCQBHIVMGVHV-UHFFFAOYSA-N
SMILESOCC(O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4700 to 1.4750 (20°C, 589 nm)
GC>=99.0 %
Water=<0.5 % (K.F.)
Sulfated ash=<0.01 %
Chloride (Cl)=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only