2-Methyl-3-buten-2-ol, 97%
2-Methyl-3-buten-2-ol, 97%
2-Methyl-3-buten-2-ol, 97%
Thermo Scientific Chemicals

2-Methyl-3-buten-2-ol, 97%

CAS: 115-18-4 | C5H10O | 86.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number 159672500
Price (USD)
34.20
Each
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
34.20
Each
Chemical Identifiers
CAS115-18-4
IUPAC Name2-methylbut-3-en-2-ol
Molecular FormulaC5H10O
InChI KeyHNVRRHSXBLFLIG-UHFFFAOYSA-N
SMILESCC(C)(O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4160 to 1.4180 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=96.0 %
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only