N,N-Dimethylbenzylamine, 99%
N,N-Dimethylbenzylamine, 99%
N,N-Dimethylbenzylamine, 99%
Thermo Scientific Chemicals

N,N-Dimethylbenzylamine, 99%

CAS: 103-83-3 | C9H13N | 135.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2.5 L
5 L
Packaging:
View all
Plastic drum
Catalog number 159750051
Price (EUR)
414,65
Online exclusive
460,00
Save 45,35 (10%)
Each
Quantity:
5 L
Packaging:
Plastic drum
Request bulk or custom format
Price (EUR)
414,65
Online exclusive
460,00
Save 45,35 (10%)
Each
Chemical Identifiers
CAS103-83-3
IUPAC Namebenzyldimethylamine
Molecular FormulaC9H13N
InChI KeyXXBDWLFCJWSEKW-UHFFFAOYSA-N
SMILESCN(C)CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.5000 to 1.5020 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Titration with HClO498.5 to 101.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only