Ethoxyacetic acid, 98%
Ethoxyacetic acid, 98%
Ethoxyacetic acid, 98%
Thermo Scientific Chemicals

Ethoxyacetic acid, 98%

CAS: 627-03-2 | C4H8O3 | 104.11 g/mol
数量:
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 159861000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS627-03-2
IUPAC Name2-ethoxyacetic acid
Molecular FormulaC4H8O3
InChI KeyYZGQDNOIGFBYKF-UHFFFAOYSA-N
SMILESCCOCC(O)=O
さらに表示
GC>=97.5 %
Refractive index1.4170 to 1.4200 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only