L(-)-Borneol, 97%
L(-)-Borneol, 97%
L(-)-Borneol, 97%
Thermo Scientific Chemicals

L(-)-Borneol, 97%

CAS: 464-45-9 | C10H18O | 154.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 160651000
Price (TWD)
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS464-45-9
IUPAC Name(1S,2R,4R)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-ol
Molecular FormulaC10H18O
InChI KeyDTGKSKDOIYIVQL-NQMVMOMDSA-N
SMILESCC1(C)[C@@H]2CC[C@]1(C)[C@H](O)C2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder or crystals
Specific optical rotation-34.7° to -37.7° (20°C, 589 nm) (c=5, C2H5OH)
Infrared spectrumConforms
Melting point204°C to 208°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only