trans-Methyl crotonate, 96%
trans-Methyl crotonate, 96%
trans-Methyl crotonate, 96%
Thermo Scientific Chemicals

trans-Methyl crotonate, 96%

CAS: 623-43-8 | C5H8O2 | 100.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 160845000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS623-43-8
IUPAC Namemethyl (2E)-but-2-enoate
Molecular FormulaC5H8O2
InChI KeyMCVVUJPXSBQTRZ-ONEGZZNKSA-N
SMILESCOC(=O)\C=C\C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4230 to 1.4270 (20°C, 589 nm)
GC>=95.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only