N,N-Dimethylformamide dimethyl acetal, ca. 97%
N,N-Dimethylformamide dimethyl acetal, ca. 97%
N,N-Dimethylformamide dimethyl acetal, ca. 97%
Thermo Scientific Chemicals

N,N-Dimethylformamide dimethyl acetal, ca. 97%

CAS: 4637-24-5 | C5H13NO2 | 119.16 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
產品號碼 161250250
價格 (TWD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS4637-24-5
IUPAC Name(dimethoxymethyl)dimethylamine
Molecular FormulaC5H13NO2
InChI KeyZSXGLVDWWRXATF-UHFFFAOYSA-N
SMILESCOC(OC)N(C)C
檢視更多
規格Specification Sheet規格表
Appearance (Color)Clear colorless
Infrared spectrumConforms
GC>=96.0 %
Refractive index1.3950 to 1.3970 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
RUO – Research Use Only