Mesitylene, 97%, pure
Mesitylene, 97%, pure
Mesitylene, 97%, pure
Thermo Scientific Chemicals

Mesitylene, 97%, pure

CAS: 108-67-8 | C9H12 | 120.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 161320010
Price (USD)
215.65
Special offer
Online exclusive
Ends: 28-Dec-2026
254.00
Save 38.35
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
215.65
Special offer
Online exclusive
Ends: 28-Dec-2026
254.00
Save 38.35
Each
Chemical Identifiers
CAS108-67-8
IUPAC Name1,3,5-trimethylbenzene
Molecular FormulaC9H12
InChI KeyAUHZEENZYGFFBQ-UHFFFAOYSA-N
SMILESCC1=CC(C)=CC(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=96.0 %
Refractive index1.4970 to 1.5010 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only