4-tert-Butylpyridine
Thermo Scientific Chemicals

4-tert-Butylpyridine

CAS: 3978-81-2 | C9H13N | 135.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 161940250
Price (TWD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS3978-81-2
IUPAC Name4-tert-butylpyridine
Molecular FormulaC9H13N
InChI KeyYSHMQTRICHYLGF-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4940 to 1.4960 (20°C, 589 nm)
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only