2,4,6-Collidine, 99%
2,4,6-Collidine, 99%
2,4,6-Collidine, 99%
Thermo Scientific Chemicals

2,4,6-Collidine, 99%

CAS: 108-75-8 | C8H11N | 121.18 g/mol
数量:
100 mL
500 mL
1 L
2.5 L
製品番号(カタログ番号) 161950025
価格(JPY)
-
数量:
2.5 L
一括またはカスタム形式をリクエストする
化学物質識別子
CAS108-75-8
IUPAC Name2,4,6-trimethylpyridine
Molecular FormulaC8H11N
InChI KeyBWZVCCNYKMEVEX-UHFFFAOYSA-N
SMILESCC1=CC(C)=NC(C)=C1
さらに表示
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4970 to 1.4990 (20°C, 589 nm)
Appearance (Form)Liquid
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only