2,6-Pyridinedicarboxylic acid chloride, 97%
2,6-Pyridinedicarboxylic acid chloride, 97%
2,6-Pyridinedicarboxylic acid chloride, 97%
Thermo Scientific Chemicals

2,6-Pyridinedicarboxylic acid chloride, 97%

CAS: 3739-94-4 | C7H3Cl2NO2 | 204.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 162131000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3739-94-4
IUPAC Namepyridine-2,6-dicarbonyl dichloride
Molecular FormulaC7H3Cl2NO2
InChI KeyGWHOGODUVLQCEB-UHFFFAOYSA-N
SMILESClC(=O)C1=CC=CC(=N1)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals and/or chunks
Appearance (Color)White to brown
Infrared spectrumConforms
Titration RCOCl>=96.0 %
Melting point57°C to 61°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only