N-Phenylanthranilic acid, 98%
N-Phenylanthranilic acid, 98%
N-Phenylanthranilic acid, 98%
Thermo Scientific Chemicals

N-Phenylanthranilic acid, 98%

CAS: 91-40-7 | C13H11NO2 | 213.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
500 g
Packaging:
Plastic bottle
Catalog number 163001000
Price (EUR)
98,40
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
98,40
Each
Chemical Identifiers
CAS91-40-7
IUPAC Name2-(phenylamino)benzoic acid
Molecular FormulaC13H11NO2
InChI KeyZWJINEZUASEZBH-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC=C1NC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point182°C to 187°C
Appearance (Color)Yellow-green to grey-green
Infrared spectrumConforms
Titration with NaOH>=97.5 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only