DL-Tyrosine, 99%
DL-Tyrosine, 99%
DL-Tyrosine, 99%
Thermo Scientific Chemicals

DL-Tyrosine, 99%

数量:
25 g
100 g
パッケージ:
Glass Bottle
製品番号(カタログ番号) 163570250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS556-03-6
IUPAC Name2-amino-3-(4-hydroxyphenyl)propanoic acid
Molecular FormulaC9H11NO3
InChI KeyOUYCCCASQSFEME-UHFFFAOYNA-N
SMILESNC(CC1=CC=C(O)C=C1)C(O)=O
さらに表示
Appearance (Form)Powder
Titration with HClO498.5 to 101.5 %
Appearance (Color)White to off-white
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only