N-Ethyl-3-carbazolecarboxaldehyde, 94%, Tech.
N-Ethyl-3-carbazolecarboxaldehyde, 94%, Tech.
N-Ethyl-3-carbazolecarboxaldehyde, 94%, Tech.
Thermo Scientific Chemicals

N-Ethyl-3-carbazolecarboxaldehyde, 94%, Tech.

CAS: 7570-45-8 | C15H13NO | 223.28 g/mol
数量:
25 g
100 g
500 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 166035000
価格(JPY)
-
数量:
500 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7570-45-8
IUPAC Name9-ethyl-9H-carbazole-3-carbaldehyde
Molecular FormulaC15H13NO
InChI KeyQGJXVBICNCIWEL-UHFFFAOYSA-N
SMILESCCN1C2=C(C=CC=C2)C2=C1C=CC(C=O)=C2
さらに表示
Appearance (Form)Crystalline powder or granules
Infrared spectrumConforms
Appearance (Color)Yellow to ochre
Melting point85°C to 91°C
HPLC>=92 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only