L-Isoleucine, 99%
L-Isoleucine, 99%
L-Isoleucine, 99%
Thermo Scientific Chemicals

L-Isoleucine, 99%

CAS: 73-32-5 | C6H13NO2 | 131.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Packaging:
View all
Plastic Bottle
Catalog number 166171000
Price (USD)
176.00
Each
Quantity:
100 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)
176.00
Each
Chemical Identifiers
CAS73-32-5
IUPAC Name(2S,3S)-2-amino-3-methylpentanoic acid
Molecular FormulaC6H13NO2
InChI KeyAGPKZVBTJJNPAG-WHFBIAKZSA-N
SMILESCC[C@H](C)[C@H](N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Specific optical rotation+38.9° to +41.8° (20°C, 589 nm) (c=4, 6 N HCl)
Iron (Fe)=<30 ppm
Heavy metals (as Pb)=<20 ppm
Appearance (Color)White to off-white
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only