2-Methylcyclohexanol, 99%, mixture of cis and trans
2-Methylcyclohexanol, 99%, mixture of cis and trans
2-Methylcyclohexanol, 99%, mixture of cis and trans
Acros Organics

2-Methylcyclohexanol, 99%, mixture of cis and trans

CAS: 583-59-5 | C7H14O | 114.19 g/mol
Have Questions?
Catalog NumberQuantity
166892500
also known as 16689
250 g
Catalog number 166892500
also known as 16689
HSN 29061200
Price (INR)
7,175.00
Each
Quantity:
250 g
Request bulk or custom format
Price (INR)
7,175.00
Each
Chemical Identifiers
CAS583-59-5
IUPAC Name2-methylcyclohexan-1-ol
Molecular FormulaC7H14O
InChI KeyNDVWOBYBJYUSMF-UHFFFAOYNA-N
SMILESCC1CCCCC1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4610 to 1.4650 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only