Decanoic acid, 99%
Decanoic acid, 99%
Decanoic acid, 99%
Thermo Scientific Chemicals

Decanoic acid, 99%

CAS: 334-48-5 | C10H20O2 | 172.268 g/mol
数量:
100 g
500 g
1 kg
2.5 kg
パッケージ:
すべて表示
Glass bottle
製品番号(カタログ番号) 167275000
価格(JPY)
-
数量:
500 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS334-48-5
IUPAC Namedecanoic acid
Molecular FormulaC10H20O2
InChI KeyGHVNFZFCNZKVNT-UHFFFAOYSA-N
SMILESCCCCCCCCCC(O)=O
さらに表示
Appearance (Form)Crystals and/or chunks or fused solid or liquid
Melting point29°C to 34°C
Infrared spectrumConforms
Appearance (Color)Colorless or white to cream
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only