1,1'-Thiocarbonyldiimidazole, 90%, Tech.
1,1'-Thiocarbonyldiimidazole, 90%, Tech.
1,1'-Thiocarbonyldiimidazole, 90%, Tech.
Thermo Scientific Chemicals

1,1'-Thiocarbonyldiimidazole, 90%, Tech.

CAS: 6160-65-2 | C7H6N4S | 178.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
Packaging:
Glass bottle
Catalog number 168480100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6160-65-2
IUPAC Name1-(1H-imidazole-1-carbothioyl)-1H-imidazole
Molecular FormulaC7H6N4S
InChI KeyRAFNCPHFRHZCPS-UHFFFAOYSA-N
SMILESS=C(N1C=CN=C1)N1C=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with HClO4>=88.0 %
Appearance (Color)Bright yellow
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only