Phosphorus tribromide, 99%
Phosphorus tribromide, 99%
Phosphorus tribromide, 99%
Thermo Scientific Chemicals

Phosphorus tribromide, 99%

CAS: 7789-60-8 | Br3P | 270.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 169475000
Price (EUR)
248,00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
248,00
Each
Chemical Identifiers
CAS7789-60-8
IUPAC Nametribromophosphane
Molecular FormulaBr3P
InChI KeyIPNPIHIZVLFAFP-UHFFFAOYSA-N
SMILESBrP(Br)Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear to slightly hazy colorless to yellow to
Appearance (Color)light brown
Appearance (Form)Liquid
Titration Argentometric98.5 to 102.0 % (ex Br)
Specific gravity2.84 to 2.89
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only