Acetic acid, ammonium salt, 98%, pure
Acetic acid, ammonium salt, 98%, pure
Acetic acid, ammonium salt, 98%, pure
Thermo Scientific Chemicals

Acetic acid, ammonium salt, 98%, pure

CAS: 631-61-8 | C2H7NO2 | 77.08 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
檢視全部
Plastic Bottle
產品號碼 169880025
價格 (TWD)
-
Quantity:
2.5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
化學識別
CAS631-61-8
IUPAC Nameammonium acetate
Molecular FormulaC2H7NO2
InChI KeyUSFZMSVCRYTOJT-UHFFFAOYSA-N
SMILES[NH4+].CC([O-])=O
檢視更多
規格Specification Sheet規格表
Appearance (Form)Adhering crystals
Titration with HClO4>=97.5 % (On anhydrous substance)
Water=<2 %
Appearance (Color)White
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only