alpha-D(+)-Glucose, 99+%, anhydrous
alpha-D(+)-Glucose, 99+%, anhydrous
alpha-D(+)-Glucose, 99+%, anhydrous
Thermo Scientific Chemicals

alpha-D(+)-Glucose, 99+%, anhydrous

CAS: 492-62-6 | C6H12O6 | 180.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
25 kg
Catalog number 170080025
Price (USD)
58.30
Each
Quantity:
2.5 kg
Request bulk or custom format
Price (USD)
58.30
Each
Chemical Identifiers
CAS492-62-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Specific optical rotationon anhydrous substance
Appearance (Color)White to almost white
Specific optical rotation+52.5° to +53.3° (20°C, 589 nm) (c=10, H2O/diluteNH3)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only