4,4'-Dimethoxybenzil, 99+%
4,4'-Dimethoxybenzil, 99+%
4,4'-Dimethoxybenzil, 99+%
Thermo Scientific Chemicals

4,4'-Dimethoxybenzil, 99+%

CAS: 1226-42-2 | C16H14O4 | 270.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 170450250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1226-42-2
IUPAC Namebis(4-methoxyphenyl)ethane-1,2-dione
Molecular FormulaC16H14O4
InChI KeyYNANGXWUZWWFKX-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1)C(=O)C(=O)C1=CC=C(OC)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point131°C to 134°C
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only