1-Phenyl-1-cyclopropanecarboxylic acid, 97%
1-Phenyl-1-cyclopropanecarboxylic acid, 97%
1-Phenyl-1-cyclopropanecarboxylic acid, 97%
Thermo Scientific Chemicals

1-Phenyl-1-cyclopropanecarboxylic acid, 97%

CAS: 6120-95-2 | C10H10O2 | 162.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 170680100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6120-95-2
IUPAC Name1-phenylcyclopropane-1-carboxylic acid
Molecular FormulaC10H10O2
InChI KeyIWWCCNVRNHTGLV-UHFFFAOYSA-N
SMILESOC(=O)C1(CC1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Infrared spectrumConforms
Melting point84°C to 87°C
GC>=96.0 %
Appearance (Form)Crystalline powder or crystals and/or chunks
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only