1-Phenyl-1-propyne, 99%
1-Phenyl-1-propyne, 99%
1-Phenyl-1-propyne, 99%
Thermo Scientific Chemicals

1-Phenyl-1-propyne, 99%

CAS: 673-32-5 | C9H8 | 116.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 171320050
Price (USD)
70.65
Online exclusive
77.90
Save 7.25
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
70.65
Online exclusive
77.90
Save 7.25
Each
Chemical Identifiers
CAS673-32-5
IUPAC Name(prop-1-yn-1-yl)benzene
Molecular FormulaC9H8
InChI KeyGHUURDQYRGVEHX-UHFFFAOYSA-N
SMILESCC#CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.5630 to 1.5650 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only