Celite™, for analysis, high purity analytical grade
Celite™, for analysis, high purity analytical grade
Celite™, for analysis, high purity analytical grade
Thermo Scientific Chemicals

Celite™, for analysis, high purity analytical grade

CAS: 61790-53-2 | O2Si | 60.083 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
Packaging:
Glass Bottle
Catalog number 173460250
Price (TWD)
-
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS61790-53-2
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Silicon dioxide (SiO2)98.7 % (typical)
Iron(III)oxide (Fe2O3)0.3 % (typical)
Calcium oxide (CaO)0.1 % (typical)
Wet density=<0.25 g/ml (centrifuged wet density) (typical)
Acid soluble iron=<5 ppm (typical)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Filter aid
RUO – Research Use Only