Thermo Scientific Chemicals

5-Nitro-2-furaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: 173860100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

5-Nitro-2-furaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: 173860100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 173860100
Price (USD)
Price: 75.80
Online price: 65.65
Your price:
Quantity
-

Chemical Identifiers

CAS
698-63-5
IUPAC Name
5-nitrofuran-2-carbaldehyde
Molecular Formula
C5H3NO4
InChI Key
SXINBFXPADXIEY-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=C(O1)C=O
Appearance (Color)
Yellow to brown
Infrared spectrum
Conforms
GC
>=96.0 %
Appearance (Form)
Low melting crystalline solid
Melting point
33°C to 37°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text