n-Amylamine, 99%
n-Amylamine, 99%
n-Amylamine, 99%
Thermo Scientific Chemicals

n-Amylamine, 99%

CAS: 110-58-7 | C5H13N | 87.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
50 mL
250 mL
Packaging:
Glass bottle
Catalog number 173940050
Price (USD)
24.65
Special offer
Online exclusive
Ends: 28-Dec-2026
29.00
Save 4.35
Each
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
24.65
Special offer
Online exclusive
Ends: 28-Dec-2026
29.00
Save 4.35
Each
Chemical Identifiers
CAS110-58-7
IUPAC Namepentan-1-amine
Molecular FormulaC5H13N
InChI KeyDPBLXKKOBLCELK-UHFFFAOYSA-N
SMILESCCCCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
AppearanceMay darken on storage
Water=<1 %
Infrared spectrumConforms
Refractive index1.4100 to 1.4120 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only